2,3,4-Trimethoxy-6-methylbenzaldehyde 95%

Supplier: Ambeed
Warning

A374403-1G A374403-250MG
76803-646EA 72 USD
76803-646 76803-648
2,3,4-Trimethoxy-6-methylbenzaldehyde 95%
2,3,4-Trimethoxy-6-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₄
MDL Number: MFCD00016598
CAS Number: 22383-85-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR