Ethyl imidazo[1,2-a]pyrazine-2-carboxylate 97%

Supplier: Ambeed
Warning

A373542-1G A373542-250MG A373542-5G
76803-636EA 76.39 USD
76803-636 76803-638 76803-640
Ethyl imidazo[1,2-a]pyrazine-2-carboxylate 97%
Ethyl imidazo[1,2-a]pyrazine-2-carboxylate

Precaución: for Research Use Only

Formula: C₉H₉N₃O₂
MW: 191.1867 g/mol
MDL Number: MFCD08460079
CAS Number: 77112-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR