Oxalic acid mono(N-methyl)amide 95%
Supplier: Ambeed
|
Danger
|
A373453-1G
A373453-5G
A373453-25G
A373453-250MG
76803-624EA
19
USD
76803-624
76803-626
77668-738
77879-814
Oxalic acid mono(N-methyl)amide 95%
Oxalic acid mono(N-methyl)amide
Precaución: for Research Use Only
Formula:
C₃H₅NO₃ |
MDL Number:
MFCD06245498 CAS Number: 29262-58-6 UN: 3077 ADR: 9,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...