5-(4-Chlorophenyl)furfural 97%

Supplier: Ambeed
Warning

Synonyms: 5-(4-Chlorophenyl)furan-2-carbaldehyde, 5-(4-Chlorophenyl)-2-furaldehyde

A371352-1G A371352-25G A371352-5G
76803-602EA 8.49 USD
76803-602 76803-604 76803-606
5-(4-Chlorophenyl)furfural 97%
5-(4-Chlorophenyl)furfural

Precaución: for Research Use Only

Formula: C₁₁H₇ClO₂
MW: 206.625 g/mol
MDL Number: MFCD00195947
CAS Number: 34035-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR