Hydridotetrakis(triphenylphosphine)rhodium(I) 98%

Supplier: Ambeed
Warning

Synonyms: Tetrakis(triphenylphosphine)rhodium(I) Hydride

A369800-100MG A369800-1G A369800-250MG
76803-580EA 59 USD
76803-580 76803-582 76803-584
Hydridotetrakis(triphenylphosphine)rhodium(I) 98%
Hydridotetrakis(triphenylphosphine)rhodium(I)

Precaución: for Research Use Only

Formula: C₇₂H₆₁P₄Rh
MW: 1153.08 g/mol
MDL Number: MFCD00015867
CAS Number: 18284-36-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR