Diethoxymethylvinylsilane 95% stabilised

Supplier: Ambeed
Danger

Synonyms: Diethoxy(methyl)(vinyl)silane

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A366026-25G A366026-5G
76803-544EA 10 USD
76803-544 76803-546
Diethoxymethylvinylsilane 95% stabilised
Diethoxymethylvinylsilane

Precaución: for Research Use Only

Formula: C₇H₁₆O₂Si
MW: 160.29 g/mol
MDL Number: MFCD00009067
CAS Number: 5507-44-8
UN: 1993
ADR: 3 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR