Ethyl-4-[(chloroacetyl)amino]benzoate 97%

Supplier: Ambeed
Warning

A365987-1G A365987-25G A365987-5G
76803-538EA 16 USD
76803-538 76803-540 76803-542
Ethyl-4-[(chloroacetyl)amino]benzoate 97%
Ethyl-4-[(chloroacetyl)amino]benzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₂ClNO₃
MDL Number: MFCD00018909
CAS Number: 26226-72-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR