2,3,4,4'-Tetrahydroxybenzophenone 98%

Supplier: Ambeed
Warning

Synonyms: (4-Hydroxyphenyl)(2,3,4-trihydroxyphenyl)methanone

A365672-100G A365672-10G A365672-25G A365672-500G A365672-5G
76803-522EA 100.79 USD
76803-522 76803-524 76803-526 76803-528 76803-530
2,3,4,4'-Tetrahydroxybenzophenone 98%
2,3,4,4'-Tetrahydroxybenzophenone

Precaución: for Research Use Only

Formula: C₁₃H₁₀O₅
MW: 246.2155 g/mol
MDL Number: MFCD00083097
CAS Number: 31127-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR