7-Isopropyl-1,4-dimethylazulene 97%

Supplier: Ambeed
Warning

Synonyms: Guaiazulene

A365642-100G A365642-1G A365642-5G A365642-25G
76803-516EA 211 USD
76803-516 76803-518 76803-520 77572-084
7-Isopropyl-1,4-dimethylazulene 97%
7-Isopropyl-1,4-dimethylazulene

Precaución: for Research Use Only

Formula: C₁₅H₁₈
MW: 198.31 g/mol
Boiling Pt: 309
Melting Pt: 133
MDL Number: MFCD00003811
CAS Number: 489-84-9
Merck Index: 12,04581

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR