(±)-Camphorquinone 98%

Supplier: Ambeed
Warning

Synonyms: 1,7,7-Trimethylbicyclo[2.2.1]heptane-2,3-dione, DL-Camphoroquinone

A365615-100G A365615-1G A365615-25G A365615-5G
76803-508EA 283 USD
76803-508 76803-510 76803-512 76803-514
(±)-Camphorquinone 98%
(±)-Camphorquinone

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₂
MW: 166.22 g/mol
MDL Number: MFCD00064160
CAS Number: 10373-78-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR