(R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(3-chlorophenyl)propanoic acid 95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-D-β-Phe(3-Cl)-OH

A365567-250MG A365567-1G
76803-506EA 32.89 USD
76803-506 77572-082
(R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(3-chlorophenyl)propanoic acid 95%
(R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(3-chlorophenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₂₄H₂₀ClNO₄
MW: 421.87 g/mol
MDL Number: MFCD03428035
CAS Number: 511272-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR