GMBS (N-Succinimidyl 4-maleimidobutyrate) 98%

Supplier: Ambeed
Warning

Synonyms: N-(γ-Maleimidobutyryloxy)succinimide, 2,5-Dioxopyrrolidin-1-yl 4-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)butanoate, N-(4-Maleimidobutyryloxy)succinimide, GMBS, 4-Maleimidobutyric acid N-hydroxysuccinimide ester, 4-Maleimidobutyric acid N-succinimidyl ester

A363934-1G A363934-250MG A363934-5G A363934-100MG A363934-25G
76803-494EA 63 USD
76803-494 76803-496 76803-498 77601-610 77879-782
GMBS (N-Succinimidyl 4-maleimidobutyrate) 98%
GMBS (N-Succinimidyl 4-maleimidobutyrate)

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂O₆
MDL Number: MFCD00036817
CAS Number: 80307-12-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR