Didecylamine 98%

Supplier: Ambeed
Warning

A363177-100G A363177-1G A363177-25G A363177-5G
76803-486EA 381.93 USD
76803-486 76803-488 76803-490 76803-492
Didecylamine 98%
Didecylamine

Precaución: for Research Use Only

Formula: C₂₀H₄₃N
MW: 297.57 g/mol
MDL Number: MFCD00048396
CAS Number: 1120-49-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR