Ethyl-N-phenylformimidate 97%
Supplier: Ambeed
|
Warning
|
A361887-1G
A361887-25G
A361887-5G
76803-474EA
6
USD
76803-474
76803-476
76803-478
Ethyl-N-phenylformimidate 97%
Ethyl-N-phenylformimidate
Precaución: for Research Use Only
Formula:
C₉H₁₁NO |
MDL Number:
MFCD00043655 CAS Number: 6780-49-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...