(2S)-2-Amino-2-(3-chlorophenyl)acetic acid 97%

Supplier: Ambeed
Warning

Synonyms: H-Phg(3-Cl)-OH, (S)-2-Amino-2-(3-chlorophenyl)acetic acid

A359542-100MG A359542-1G A359542-250MG
76803-450EA 7 USD
76803-450 76803-452 76803-454
(2S)-2-Amino-2-(3-chlorophenyl)acetic acid 97%
(2S)-2-Amino-2-(3-chlorophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₈H₈ClNO₂
MDL Number: MFCD03839814
CAS Number: 119565-00-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR