8-hydroxyquinoline-2-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 8-Hydroxyquinoline-2-carboxylic acid

A359083-100MG A359083-1G A359083-250MG A359083-5G
76803-434EA 28.65 USD
76803-434 76803-436 76803-438 76803-440
8-hydroxyquinoline-2-carboxylic acid 95%
8-hydroxyquinoline-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₇NO₃
MW: 189.17 g/mol
MDL Number: MFCD00168972
CAS Number: 1571-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR