Glycerol formal (mixture of 1,3-dioxan-5-ol and 4-hydroxymethyldioxolane) 98% mix

Supplier: Ambeed
Warning

Synonyms: Glycerol Formal (mixture of 1,3-Dioxan-5-ol and 4-Hydroxymethyldioxolane)

A358784-1000G A358784-100G A358784-25G A358784-500G
76803-424EA 40.32 USD
76803-424 76803-426 76803-428 76803-430
Glycerol formal (mixture of 1,3-dioxan-5-ol and 4-hydroxymethyldioxolane) 98% mix
Glycerol formal (mixture of 1,3-dioxan-5-ol and 4-hydroxymethyldioxolane)

Precaución: for Research Use Only

Formula: C₄H₈O₃
MW: 104.11 g/mol
MDL Number: MFCD00003218
CAS Number: 99569-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR