Acetaldehyde ammonia trimer 95% (by titrimetric analysis)

Supplier: Ambeed
Warning

Synonyms: Aldehyde Ammonia Trimer, 2,4,6-Trimethylhexahydro-1,3,5-triazine Trihydrate, Hexahydro-2,4,6-trimethyl-1,3,5-triazine Trihydrate, 2,4,6-trimethyl-1,3,5-triazinane trihydrate

A357084-25G A357084-5G
76803-406EA 37.14 USD
76803-406 76803-408
Acetaldehyde ammonia trimer 95% (by titrimetric analysis)
Acetaldehyde ammonia trimer

Precaución: for Research Use Only

Formula: C₆H₂₁N₃O₃
MW: 183.25 g/mol
MDL Number: MFCD00149559
CAS Number: 58052-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR