Magnesium carbonate heavy 99.99%

Supplier: Ambeed
Danger

A355376-1000G A355376-100G A355376-25G A355376-500G
76803-374EA 20 USD
76803-374 76803-376 76803-378 76803-380
Magnesium carbonate heavy 99.99%
Magnesium carbonate heavy

Precaución: for Research Use Only

Formula: CH₂Mg₂O₅
MDL Number: MFCD00011107
CAS Number: 39409-82-0
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR