Ethyl-2,4-dichloropyrimidine-5-carboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 2,4-Dichloro-5-pyrimidinecarboxylate
A354856-100G
A354856-10G
A354856-1G
A354856-25G
A354856-500G
A354856-5G
76803-362EA
240.83
USD
76803-362
76803-364
76803-366
76803-368
76803-370
76803-372
Ethyl-2,4-dichloropyrimidine-5-carboxylate 97%
Ethyl-2,4-dichloropyrimidine-5-carboxylate
Precaución: for Research Use Only
Formula:
C₇H₆Cl₂N₂O₂ MW: 221.04 g/mol |
MDL Number:
MFCD09910281 CAS Number: 51940-64-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...