4'-(Trifluoromethyl)propiophenone 97%

Supplier: Ambeed
Warning

Synonyms: p-(Trifluoromethyl)propiophenone, 1-(4-(Trifluoromethyl)phenyl)propan-1-one

A354645-100G A354645-10G A354645-1G A354645-25G A354645-5G
76803-352EA 334.19 USD
76803-352 76803-354 76803-356 76803-358 76803-360
4'-(Trifluoromethyl)propiophenone 97%
4'-(Trifluoromethyl)propiophenone

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O
MW: 202.18 g/mol
MDL Number: MFCD00039231
CAS Number: 711-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR