4'-Fluoro-biphenyl-4-ylamine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4'-Fluoro-[1,1'-biphenyl]-4-amine
A354205-100MG
A354205-1G
A354205-250MG
A354205-5G
76803-344EA
41.38
USD
76803-344
76803-346
76803-348
76803-350
4'-Fluoro-biphenyl-4-ylamine 98%
4'-Fluoro-biphenyl-4-ylamine
Precaución: for Research Use Only
Formula:
C₁₂H₁₀FN MW: 187.22 g/mol |
MDL Number:
MFCD00025338 CAS Number: 324-93-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...