Fmoc-α-methyl-L-phenylalanine 95%

Supplier: Ambeed
Warning

Synonyms: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-methyl-3-phenylpropanoic acid, Fmoc-α-Me-Phe-OH

A354098-100MG A354098-5G A354098-25G A354098-1G
76803-338EA 16 USD
76803-338 77568-714 77572-040 77879-764
Fmoc-α-methyl-L-phenylalanine 95%
Fmoc-alpha-methyl-L-phenylalanine

Precaución: For research use only.

Formula: C₂₅H₂₃NO₄
MW: 401.47 g/mol
MDL Number: MFCD02682285
CAS Number: 135944-05-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR