Marbofloxacin 98+%

Supplier: Ambeed
Warning

Synonyms: 9-Fluoro-3,7-dihydro-3-methyl-10-(4-methyl-1-piperazinyl)-7-oxo-2H-[1,3,4]oxadiazino[6,5,4-ij]quinoline-6-carboxylic acid, 9-Fluoro-3-methyl-10-(4-methylpiperazin-1-yl)-7-oxo-3,7-dihydro-2H-[1,3,4]oxadiazino[6,5,4-ij]quinoline-6-carboxylic acid

A352968-100G A352968-100MG A352968-1G A352968-250MG A352968-25G A352968-25MG A352968-5G A352968-1MG
76803-314EA 1076.82 USD
76803-314 76803-316 76803-318 76803-320 76803-322 76803-324 76803-326 77668-638
Marbofloxacin 98+%
Marbofloxacin

Precaución: for Research Use Only

Formula: C₁₇H₁₉FN₄O₄
MW: 362.36 g/mol
MDL Number: MFCD00864820
CAS Number: 115550-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR