Isophthalic dihydrazide 97%

Supplier: Ambeed
Warning

A352388-25G A352388-500G
76803-306EA 23 USD
76803-306 76803-308
Isophthalic dihydrazide 97%
Isophthalic dihydrazide

Precaución: for Research Use Only

Formula: C₈H₁₀N₄O₂
MDL Number: MFCD00025117
CAS Number: 2760-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR