Elacridar 98%

Supplier: Ambeed
Warning

Synonyms: N-[4-[2-(3,4-Dihydro-6,7-dimethoxy-2(1H)-isoquinolinyl)ethyl]phenyl]-9,10-dihydro-5-methoxy-9-oxo-4-acridinecarboxamide, N-(4-(2-(6,7-Dimethoxy-3,4-dihydroisoquinolin-2(1H)-yl)ethyl)phenyl)-5-methoxy-9-oxo-9,10-dihydroacridine-4-carboxamide

A351095-1G A351095-1MG A351095-5MG A351095-10MG A351095-100MG A351095-250MG A351095-50MG
76803-294EA 765.97 USD
76803-294 76803-296 76803-298 77601-570 77626-672 77626-674 77626-676
Elacridar 98%
Elacridar

Precaución: for Research Use Only

Formula: C₃₄H₃₃N₃O₅
MW: 563.654 g/mol
MDL Number: MFCD00912604
CAS Number: 143664-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR