Diethyl cyclopentane-1,1-dicarboxylate 97%

Supplier: Ambeed
Warning

A350634-10G A350634-1G A350634-250MG A350634-5G
76803-286EA 70 USD
76803-286 76803-288 76803-290 76803-292
Diethyl cyclopentane-1,1-dicarboxylate 97%
Diethyl cyclopentane-1,1-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₈O₄
MDL Number: MFCD00219321
CAS Number: 4167-77-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR