1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic acid 95%

Supplier: Ambeed
Warning

Synonyms: DOTA, 1,4,7,10-Tetraazacyclododecane-N,N',N'',N'''-tetraacetic acid, 2,2',2'',2'''-(1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetrayl)tetraacetic acid

A349340-100G A349340-10G A349340-1G A349340-250MG A349340-25G A349340-5G
76803-248EA 712.93 USD
76803-248 76803-250 76803-252 76803-254 76803-256 76803-258
1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic acid 95%
1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic acid

Precaución: for Research Use Only

Formula: C₁₆H₂₈N₄O₈
MW: 404.42 g/mol
MDL Number: MFCD00068657
CAS Number: 60239-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR