Diethylene glycol monoethyl ether 99%

Supplier: Ambeed
Warning

Synonyms: 2-(2-Ethoxyethoxy)ethanol, Di(ethylene glycol) ethyl ether, Diethylene Glycol Monoethyl Ether

A349114-1000G A349114-100G A349114-500G
76803-236EA 16.98 USD
76803-236 76803-238 76803-240
Diethylene glycol monoethyl ether 99%
Diethylene glycol monoethyl ether

Precaución: for Research Use Only

Formula: C₆H₁₄O₃
MW: 134.18 g/mol
MDL Number: MFCD00002872
CAS Number: 111-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR