Indan-5-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: Indane-5-carboxylic Acid

A347938-100MG A347938-1G A347938-250MG
76803-228EA 30 USD
76803-228 76803-230 76803-232
Indan-5-carboxylic acid 95%
Indan-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
MDL Number: MFCD04038659
CAS Number: 65898-38-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR