(R)-2-Amino-3-cyclohexylpropanoic acid hydrochloride 97%

Supplier: Ambeed
Warning

A347269-100G A347269-1G A347269-25G A347269-5G
76803-216EA 623.81 USD
76803-216 76803-218 76803-220 76803-222
(R)-2-Amino-3-cyclohexylpropanoic acid hydrochloride 97%
(R)-2-Amino-3-cyclohexylpropanoic acid hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₈ClNO₂
MW: 207.70 g/mol
MDL Number: MFCD00190860
CAS Number: 151899-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR