1,2,3,6-Tetrahydro-4-phenylpyridine hydrochloride 98%

Supplier: Ambeed
Danger

A347081-1G A347081-25G A347081-5G
76803-212EA 8 USD
76803-212 76803-214 77601-552
1,2,3,6-Tetrahydro-4-phenylpyridine hydrochloride 98%
1,2,3,6-Tetrahydro-4-phenylpyridine hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₃N
MDL Number: MFCD00012752
CAS Number: 43064-12-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR