bisBenzimide H 33258 trihydrochloride hydrate (Hoechst 33258) 98%

Supplier: Ambeed
Warning

Synonyms: Bisbenzimide H 33258, 4-(5-(4-Methylpiperazin-1-yl)-1H,1'H-[2,5'-bibenzo[d]imidazol]-2'-yl)phenol trihydrochloride, 2'-(4-Hydroxyphenyl)-5-(4-methyl-1-piperazinyl)-2,5'-bibenzimidazole trihydrochloride hydrate

A346521-100MG A346521-1G A346521-1MG A346521-50MG
76803-204EA 105.03 USD
76803-204 76803-206 76803-208 76803-210
bisBenzimide H 33258 trihydrochloride hydrate (Hoechst 33258) 98%
bisBenzimide H 33258 trihydrochloride hydrate (Hoechst 33258)

Precaución: for Research Use Only

Formula: C₂₅H₂₄N₆O·3HCl·xH₂O
MW: 533.88 g/mol (anhy)
MDL Number: MFCD00012679
CAS Number: 23491-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR