Genipin 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Methyl (1S,2R,6S)-2-Hydroxy-9-(hydroxymethyl)-3-oxabicyclo[4.3.0]nona-4,8-diene-5-carboxylate, (1R,4aS,7aS)-Methyl 1-hydroxy-7-(hydroxymethyl)-1,4a,5,7a-tetrahydrocyclopenta[c]pyran-4-carboxylate
A346395-100MG
A346395-1G
A346395-1MG
A346395-250MG
A346395-5G
A346395-25G
76803-194EA
6.37
USD
76803-194
76803-196
76803-198
76803-200
76803-202
77568-692
Genipin 98%
Genipin
Precaución: For research use only.
Formula:
C₁₁H₁₄O₅ MW: 226.23 g/mol |
MDL Number:
MFCD00888600 CAS Number: 6902-77-8 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...