Ethyl-2-methyl-5-nitro-3-pyridinecarboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-methyl-5-nitro-3-pyridinecarboxylate

A346390-1G A346390-25G A346390-5G
76803-188EA 51.99 USD
76803-188 76803-190 76803-192
Ethyl-2-methyl-5-nitro-3-pyridinecarboxylate 98%
Ethyl-2-methyl-5-nitro-3-pyridinecarboxylate

Precaución: for Research Use Only

Formula: C₉H₁₀N₂O₄
MW: 210.1867 g/mol
MDL Number: MFCD00753850
CAS Number: 51984-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR