meso-Erythritol 97%

Supplier: Ambeed
Warning

Synonyms: meso-Erythritol, (2R,3S)-rel-Butane-1,2,3,4-tetraol, i-Erythritol

A345952-10G A345952-25G A345952-50MG A345952-100G A345952-500G A345952-1000G
76803-182EA 6.37 USD
76803-182 76803-184 76803-186 77568-688 77568-690 77601-546
meso-Erythritol 97%
meso-Erythritol

Precaución: For research use only.

Formula: C₄H₁₀O₄
MW: 122.12 g/mol
MDL Number: MFCD00004710
CAS Number: 149-32-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR