3-Benzo[b]furylacetonitrile 98%

Supplier: Ambeed
Warning

A345120-100MG A345120-1G A345120-250MG A345120-5G
76803-172EA 8 USD
76803-172 76803-174 76803-176 76803-178
3-Benzo[b]furylacetonitrile 98%
3-Benzo[b]furylacetonitrile

Precaución: for Research Use Only

Formula: C₁₀H₇NO
MDL Number: MFCD02183555
CAS Number: 52407-43-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR