Furfuryl isopropyl sulfide 98%

Supplier: Ambeed
Warning

Synonyms: 2-((Isopropylthio)methyl)furan

A344950-100G A344950-25G A344950-5G
76803-166EA 223.85 USD
76803-166 76803-168 76803-170
Furfuryl isopropyl sulfide 98%
Furfuryl isopropyl sulphide

Precaución: for Research Use Only

Formula: C₈H₁₂OS
MW: 156.25 g/mol
MDL Number: MFCD00040265
CAS Number: 1883-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR