3-(4-Chlorophenyl)pentanedioic acid 97%

Supplier: Ambeed
Warning

A344637-1000G A344637-100G A344637-10G A344637-25G A344637-500G A344637-5G
76803-154EA 411.63 USD
76803-154 76803-156 76803-158 76803-160 76803-162 76803-164
3-(4-Chlorophenyl)pentanedioic acid 97%
3-(4-Chlorophenyl)pentanedioic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₁ClO₄
MDL Number: MFCD00190249
CAS Number: 35271-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR