(6-Methoxybenzo[b]furan-3-yl)acetic acid 97%

Supplier: Ambeed
Danger

Synonyms: 2-(6-Methoxybenzofuran-3-yl)acetic acid, 3-(Carboxymethyl)-6-methoxybenzo[b]furan

A344095-1G A344095-250MG A344095-5G
76803-148EA 56 USD
76803-148 76803-150 77879-738
(6-Methoxybenzo[b]furan-3-yl)acetic acid 97%
(6-Methoxybenzo[b]furan-3-yl)acetic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₀O₄
MDL Number: MFCD02677734
CAS Number: 69716-05-8
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR