8-Chloroquinaldine 99%

Supplier: Ambeed
Warning

Synonyms: 8-Chloro-2-methylquinoline

A343047-1G A343047-250MG A343047-5G
76803-142EA 33.95 USD
76803-142 76803-144 76803-146
8-Chloroquinaldine 99%
8-Chloroquinaldine

Precaución: for Research Use Only

Formula: C₁₀H₈ClN
MW: 177.63 g/mol
MDL Number: MFCD00051764
CAS Number: 3033-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR