Isoquinolin-1-ylmethanol 95%

Supplier: Ambeed
Warning

Synonyms: 1-(Hydroxymethyl)isoquinoline

A342824-250MG A342824-100MG
76803-130EA 99.73 USD
76803-130 77594-004
Isoquinolin-1-ylmethanol 95%
Isoquinolin-1-ylmethanol

Precaución: for Research Use Only

Formula: C₁₀H₉NO
MW: 159.1846 g/mol
MDL Number: MFCD03086097
CAS Number: 27311-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR