Dichlorodicarbonylbis(triphenylphosphine)ruthenium(II) 98%
Supplier: Ambeed
|
Warning
|
A342648-1G
A342648-250MG
A342648-5G
76803-124EA
31
USD
76803-124
76803-126
76803-128
Dichlorodicarbonylbis(triphenylphosphine)ruthenium(II) 98%
Dichlorodicarbonylbis(triphenylphosphine)ruthenium(II)
Precaución: for Research Use Only
Formula:
C₃₈H₃₄Cl₂O₂P₂Ru MW: 756.6000 g/mol |
MDL Number:
MFCD00009594 CAS Number: 14564-35-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...