7-Hydroxycoumarin-4-acetic acid 97%
Supplier: Ambeed
|
Warning
|
A340782-10G
A340782-1G
A340782-250MG
A340782-25G
A340782-5G
76803-114EA
85
USD
76803-114
76803-116
76803-118
76803-120
76803-122
7-Hydroxycoumarin-4-acetic acid 97%
7-Hydroxycoumarin-4-acetic acid
Precaución: for Research Use Only
Formula:
C₁₁H₈O₅ |
MDL Number:
MFCD00037563 CAS Number: 6950-82-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...