2-{[3-(Trifluoromethyl)benzoyl]amino}acetic acid 95%

Supplier: Ambeed
Warning

A339960-100MG A339960-250MG
76803-110EA 16 USD
76803-110 76803-112
2-{[3-(Trifluoromethyl)benzoyl]amino}acetic acid 95%
2-{[3-(Trifluoromethyl)benzoyl]amino}acetic acid

Precaución: for Research Use Only

Formula: C₁₀H₈F₃NO₃
MDL Number: MFCD00029782
CAS Number: 17794-48-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR