Iron(II) acetate 95%

Supplier: Ambeed
Warning

A338405-100G A338405-25G A338405-500G
76803-102EA 180 USD
76803-102 76803-104 76803-106 77668-554
Iron(II) acetate 95%
Iron(II) acetate

Precaución: for Research Use Only

Formula: C₄H₆FeO₄
MDL Number: MFCD00058909
CAS Number: 3094-87-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR