Manganese(II,III) oxide Mn≥71%, BET:5-7m2/g

Supplier: Ambeed
Warning

A336718-1000G A336718-100G A336718-500G
76803-084EA 26 USD
76803-084 76803-086 76803-088
Manganese(II,III) oxide Mn≥71%, BET:5-7m2/g
Manganese(II,III) oxide

Precaución: for Research Use Only

Formula: Mn₃O₄
MW: 228.81
MDL Number: MFCD00016218
CAS Number: 1317-35-7
Merck Index: 13,05753

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR