Hexaammineruthenium(II) chloride 98%

Supplier: Ambeed
Warning

A335228-100MG A335228-1G A335228-250MG
76803-076EA 41.38 USD
76803-076 76803-078 76803-080
Hexaammineruthenium(II) chloride 98%
Hexaammineruthenium (II) chloride

Precaución: for Research Use Only

Formula: H₁₈Cl₂N₆Ru
MW: 274.16 g/mol
MDL Number: MFCD00011477
CAS Number: 15305-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR