Maraviroc 99%

Supplier: Ambeed
Warning

Synonyms: UK427857, MVC, 4,4-Difluoro-N-((S)-3-((1R,3R,5S)-3-(3-isopropyl-5-methyl-4H-1,2,4-triazol-4-yl)-8-azabicyclo[3.2.1]octan-8-yl)-1-phenylpropyl)cyclohexane-1-carboxamide, 4,4-difluoro-N-[(1S)-3-[(3-exo)-3-[3-methyl-5-(1-methylethyl)-4H-1,2,4-triazol-4-yl]-8-azabicyclo[3.2.1]oct-8-yl]-1-phenylpropyl]-cyclohexanecarboxamide

A330828-1MG A330828-5MG A330828-1G
76803-054EA 11.67 USD
76803-054 76803-056 77668-494
Maraviroc 99%
Maraviroc

Precaución: for Research Use Only

Formula: C₂₉H₄₁F₂N₅O
MW: 513.67 g/mol
MDL Number: MFCD13188530
CAS Number: 376348-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR