Lopinavir 99+%

Supplier: Ambeed
Warning

Synonyms: Koletra, N-((2S,4S,5S)-5-(2-(2,6-Dimethylphenoxy)acetamido)-4-hydroxy-1,6-diphenylhexan-2-yl)-3-methyl-2-(2-oxotetrahydropyrimidin-1(2H)-yl)butanamide, ABT-378, Aluviran, A-157378.0, (S)-N-((2R,4S,5S)-5-(2-(2,6-Dimethylphenoxy)acetamido)-4-hydroxy-1,6-diphenylhexn-2-yl)-3-methyl-2-(2-oxotetrahydropyrimidin-1(2H)-yl)butanamide

A330804-100MG A330804-10MG A330804-1MG A330804-250MG A330804-50MG A330804-5G
76803-042EA 42.44 USD
76803-042 76803-044 76803-046 76803-048 76803-050 76803-052
Lopinavir 99+%
Lopinavir

Precaución: for Research Use Only

Formula: C₃₇H₄₈N₄O₅
MW: 628.82 g/mol
MDL Number: MFCD22628840
CAS Number: 192725-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR